Learn More
Medchemexpress LLC Ns2b/NS3-in-2 | 3010940-08-3 | 98.1% | 463.51 g/mol | C24H21N3O5S | 5 MG

Supplier: Medchemexpress LLC HY1446125MG
NS2B/NS3-IN-2 is a research-grade covalent inhibitor of dengue virus NS2B/NS3 protease with potent biochemical activity and reported lack of cytotoxicity in assay systems. It is supplied as a high-purity powder intended for laboratory research use only.
- Potent NS2B/NS3 protease inhibition (IC50 6.0 nM).
- Binding affinity Ki of 0.66 μM.
- High purity: 98.1%.
- Molecular formula C24H21N3O5S; molecular weight 463.51 g/mol.
- Soluble in DMSO at 100 mg/mL; in vivo formulation example 10% DMSO/90% corn oil.
- Storage: powder -20°C (up to 3 years); in solution -80°C (up to 6 months).
- For research use only; not for human or clinical use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.