Learn More
Medchemexpress LLC NS3623 | 343630-41-1 | 99.0% | 427.18 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY10858610MM/1ML
Encompass_Preferred
NS3623 is an activator of human ether-a-go-go-related gene (hERG1/KV11.1) potassium channels. It activates the IKr and Ito currents and exhibits antiarrhythmic effects. It also has a dual mode of action, acting as an inhibitor of hERG1 channels. NS3623 is suitable for laboratory research and studies related to cardiac electrophysiology.
  • Increases IKr tail currents in isolated canine cardiomyocytes
  • Shortens corrected QT interval in anesthetized guinea pigs
  • Improves repolarization reserve
  • Normalizes ventricular action potentials
  • Exhibits antiarrhythmic effects
Catalog No. 50-003-44310