Learn More
Medchemexpress LLC NS3694 | 426834-38-0 | 99.9% | 358.70 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY10835610MG
Encompass_Preferred
NS3694 is a diarylurea compound that acts as an apoptosome inhibitor. It inhibits apoptosome formation and caspase activation. It is for research use only.
  • Inhibits apoptosome formation and caspase activation.
  • Specifically suppresses the cytochrome C-induced formation of apoptosome complex and caspase-9 activation, inhibiting mitochondria-mediated apoptosis.
  • Inhibits the formation of the apoptosome Apaf-1 by blocking the activation of the initiator caspase-9.
Catalog No. 50-003-47004