Learn More
Selleck Chemical LLC NS6180 S0090-5MG
SDP
Supplier:  Selleck Chemical LLC S00905MG
Encompass_Preferred
NS-6180 is a highly selective inhibitor of KCa3 1 ion channel with IC50 value of 9 nM which inhibits T cell activation and inflammation
Catalog No. 50-000-39810