Learn More
Medchemexpress LLC NSC 107512 | 22242-89-3 | 100.0% | 324.29 | C12H16N6O5 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1416875MG
Encompass_Preferred
NSC 107512 is a sangivamycin-like small molecule inhibitor of cyclin-dependent kinase 9 (CDK9) developed for research use. It has been shown to inhibit tumor cell growth and induce apoptosis in preclinical studies, and is supplied as a solid with characterized purity and analytical data to support biochemical and cell-based assays.
  • High purity: 99.97%.
  • Targeted CDK9 inhibitory activity for biochemical and cell-based assays.
  • Supplied as a white to off-white solid for ease of handling and storage.
  • Available in small milligram quantities and ready-to-use DMSO solution formats.
  • Molecular formula and molecular weight provided to support experimental planning.
Catalog No. 50-002-21508