Learn More
Medchemexpress LLC NSC 107512 | 22242-89-3 | 99.97% | C12H16N6O5 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY14168710MM/1ML
Encompass_Preferred
NSC 107512 is a potent inhibitor of cyclin-dependent kinase 9 (CDK9). It belongs to a class of sangivamycin-like molecules (SLM). It inhibits growth and induces apoptosis of multiple myeloma tumors.
  • Potent inhibitor of cyclin-dependent kinase 9 (CDK9)
  • Belongs to a class of sangivamycin-like molecules (SLM)
  • Inhibits growth of multiple myeloma tumors
  • Induces apoptosis of multiple myeloma tumors
Catalog No. 50-003-53407