Learn More
Medchemexpress LLC NSC 146109 hydrochloride | 59474-01-0 | MFCD09878281 | 99.9% | 316.85 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY10863810MM/1ML
Encompass_Preferred
NSC 146109 hydrochloride is a small-molecule p53 activator that targets MDMX and is suitable for breast cancer research. As a pseudourea derivative, it functions by activating p53 and inducing the expression of proapoptotic genes, thereby promoting apoptosis in breast cancer cells.
  • Activates p53
  • Targets MDMX
  • Induces expression of proapoptotic genes
  • Promotes apoptosis in breast cancer cells
  • Pseudourea derivative
Catalog No. 50-003-46198