Learn More
Medchemexpress LLC NSC 194308 | 90379-42-3 | C12H22N2O3S2 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY153736
Encompass_Preferred
NSC 194308 is an enhancer of U2AF2-RNA complexes, increasing the association of the U2AF1-U2AF2-SF1-splice site RNA complex by binding between the RRM1 and RRM2 motifs of U2AF2. This compound inhibits pre-mRNA splicing by stopping spliceosome assembly when U2AF recruits U2 snRNP to the branchpoint, and also enhances pre-mRNA binding to U2AF2, leading to selective cell death in leukemia cell lines with spliceosome mutations.
  • Enhances U2AF2-RNA complexes
  • Increases association of U2AF1-U2AF2-SF1-splice site RNA complex
  • Binds between RRM1 and RRM2 motifs of U2AF2
  • Inhibits pre-mRNA splicing by stalling spliceosome assembly
  • Selectively triggers cell death in leukemia cell lines with spliceosome mutations
Catalog No. 50-003-75606