Learn More
TARGETMOL CHEMICALS INC NSC 23766 TRIHYDROCHLORI 25MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T634225MG
Encompass_Preferred
Also available in 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. NSC 23766 trihydrochloride (Rac1 Inhibitor) is an inhibitor of Rac GTPase targeting Rac activation by GEFs (IC50 ~50 uM); no inhibitory for the closely related targets RhoA or Cdc42. purity: 99%
Catalog No. 50-257-9423