Learn More
Medchemexpress LLC NSC 357754 dihydrochloride | 98.0% | 493.38 | 1 MG
Supplier: Medchemexpress LLC HY171152A1MG
NSC 357754 dihydrochloride is an inhibitor of Claudin. It can increase the transepithelial electrical resistance of cells and reduce the permeability of specific cations. It can be used in the research of intestinal diseases mediated by Claudin-2 and/or Claudin-15.
- Inhibits claudin
- Increases transepithelial electrical resistance of cells
- Reduces permeability of specific cations
- Can be used in research of intestinal diseases mediated by claudin-2 and/or claudin-15
- Appears as a solid
- Color is light yellow to yellow
- Insoluble in H2O (< 0.1 mg/mL)