Learn More
Medchemexpress LLC HY-119172 5mg Medchemexpress, NSC-60339 CAS:70-09-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1191725MG
Encompass_Preferred
Medchemexpress, HY-119172 5mg NSC-60339 CAS:70-09-7 NSC-60339, an efflux pump inhibitor and a substrate of AcrAB-TolC, is a polybasic terephthalic acid derivative studied as a potential cancer chemotherapeutic agent[2]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9848