Learn More
Apexbio Technology LLC NSC 663284 383907-43-5 100mg
SDP
Supplier:  Apexbio Technology LLC B6881100
Encompass_Preferred
NSC 663284 (CAS 383907-43-5) is a small molecule belonging to the quinolinedione class characterized as an inhibitor of Cdc25 phosphatases By targeting Cdc25 enzymes it interferes with the dephosphorylation of cyclin-dependent kinases a process essential for cell cycle progression NSC 663284 is utilized in biomedical research to study cell cycle regulation mechanisms and has potential utility in exploring therapeutic strategies aimed at modulating mitotic control in proliferative diseases
Catalog No. 50-000-92022