Learn More
Medchemexpress LLC NSC 694623 | 907957-34-0 | 99.5% | 284.38 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1472905MG
Encompass_Preferred
NSC 694623 is a potent histone acetyltransferase (HAT) inhibitor, specifically targeting recombinant HAT p300/CBP-associated factor (PCAF) with an IC50 value of 15.9 μM. It demonstrates antiproliferative activity against certain cancer cells, making it suitable for anticancer research. This compound exhibits an IC50 of 8.93 μM against SK-N-SH cells and inhibits 21% of HCT116 cells at 25 μM.
  • Potent inhibitor of histone acetyltransferase (HAT) PCAF.
  • Exhibits antiproliferative activity against specific cancer cells.
  • Useful for research applications in anticancer studies.
  • Shows growth inhibition against human SK-N-SH cells.
Catalog No. 50-003-60782