Learn More
Selleck Chemical LLC S3I-201 50mg 501919-59-1 NSC 74859
SDP
Supplier:  Selleck Chemical LLC S115550MG
Encompass_Preferred
S3I-201 (NSC 74859) shows potent inhibition of STAT3 DNA-binding activity with IC50 of 86 ?M in cell-free assays, and low activity towards STAT1 and STAT5. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-865-3