Learn More
Selleck Chemical LLC NSC2805 E5844-5MG
SDP
Supplier:  Selleck Chemical LLC E58445MG
Encompass_Preferred
NSC2805 is a potent inhibitor of WWP2 ubiquitin ligase with an IC50 of 0 38 M It is used in cancer research for studying WWP2-associated pathways
Catalog No. 50-000-42353