Learn More
Medchemexpress LLC NSC668394 25mg | 382605-72-3 | 452.10 | C17H12Br2N2O3 | 25 MG
SDP
Supplier:  Medchemexpress LLC HY11549225MG
Encompass_Preferred
NSC668394 is a small-molecule inhibitor that blocks ezrin phosphorylation at Thr567. It is supplied as a high-purity solid for research use and is commonly used in cellular and biochemical studies of ezrin-mediated signaling and metastatic behavior.
  • Inhibits ezrin Thr567 phosphorylation.
  • High purity suitable for biochemical and cellular assays.
  • Supplied as an orange to red solid for ease of handling.
  • Available in small-quantity vials for laboratory research.
  • For research use only; not for human or veterinary use.
Catalog No. 50-002-29180