Learn More
Medchemexpress LLC NSC668394 50mg | 382605-72-3 | 452.10 g·mol⁻¹ | C17H12Br2N2O3 | 50 MG

Supplier: Medchemexpress LLC HY11549250MG
NSC668394 (7-[2-(3,5-dibromo-4-hydroxyphenyl)ethylamino]quinoline-5,8-dione) is a research-grade small-molecule inhibitor of ezrin Thr567 phosphorylation used to study cell signaling and tumor metastasis mechanisms. Supplied as a solid for laboratory research use only.
- CAS number: 382605-72-3.
- Molecular formula: C17H12Br2N2O3.
- Molecular weight: 452.10 g·mol⁻¹.
- Reported purity: 99.31% (manufacturer reported).
- Target: inhibits ezrin phosphorylation at Thr567.
- Physical form: solid powder suitable for in vitro studies.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.