Learn More
Medchemexpress LLC NSC668394 5mg | 382605-72-3 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1154925MG
Encompass_Preferred
NSC668394 is a small-molecule inhibitor of ezrin phosphorylation (Thr567) used in research to probe ezrin-dependent signaling and tumor cell migration. The compound is supplied for laboratory in vitro studies and is characterized for purity and identity by the manufacturer.
  • Inhibits ezrin T567 phosphorylation; reported Kd 12.59 μM.
  • High purity, approximately 99.3%.
  • Chemical formula C17H12Br2N2O3 and molecular weight 452.10 g·mol⁻1.
  • Supplied as a small solid suitable for in vitro research applications.
Catalog No. 50-003-08109