Learn More
Medchemexpress LLC NSC693868 1MG
SDP
Supplier:  Medchemexpress LLC HY1033811MG
Encompass_Preferred
NSC693868 is a selective inhibitor of CDK1 and CDK5 with IC50s of 600 nM and 400 nM, respectively. NSC693868 less potently inhibits GSK3β with an IC50 of 1 µM) and does not block CDC25 activity. NSC693868 is used to help define the roles of CDK1 and CDK5 in various signaling pathways. cas no:40254-90-8
Catalog No. NC3440440