Learn More
TARGETMOL CHEMICALS INC NSI-189 Phosphate 50MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T2124450MG
Encompass_Preferred
Also available in 1 mL, 5 mg, 10 mg, 25 mg, 100 mg, 200 mg and bulk. Please contact Fisher for quotes.NSI-189 Phosphate is a novel neurogenic molecule with pleiotropic properties, including antidepressant, procognitive, synaptoplastic, and neurotrophic activities demonstrated in preclinical studies. Its antidepressant activity is monoamine-independent. Purity 99.95%
Catalog No. 50-287-8974