Learn More
ABclonal Technology NSUN2 Rabbit mAb
SDP
Supplier:  ABclonal Technology A2413220UL
Encompass_Preferred
This gene encodes a methyltransferase that catalyzes the methylation of cytosine to 5-methylcytosine (m5C) at position 34 of intron-containing tRNA(Leu)(CAA) precursors This modification is necessary to stabilize the anticodon-codon pairing and correctly translate the mRNA Alternatively spliced transcript variants encoding different isoforms have been noted for this gene
Catalog No. 50-000-06961