Learn More
Advanced ImmunoChemical, Inc. Recombinant Human Insulin-Like Growth Factor Binding Protein 4, N-Terminal Fragment (NT-IGFBP-4), Source - Expressed in Human Cell LinePurity > 95% (Tricine-Sds-Page)
Supplier:  Advanced ImmunoChemical, Inc. 8IGBPNRH0.05MG
Encompass_Preferred
This item is manufactured by HyTest, Ltd and distributed by Advanced ImmunoChemical, Inc. Antigens / Cardiac Markers / Recombinant human Insulin-like growth factor binding protein 4, N-terminal fragment (NT-IGFBP-4). Source:Expressed in human cell lineAnalysis:Purity > 95% (Tricine-SDS-PAGE).NT-IGFBP-4 concentration is determined spectrophotometrically.Immunoreactivity is confirmed by reaction with monoclonal antibodies specific to different epitopes of NT-IGFBP-4 molecule.Purification:Immunoaffinity purificationPresentation:Lyophilized from 20 mM Tris-HCl, 150 mM NaCl, pH 7.4Storage Conditions:Before and after reconstitution at -70 °C (between -65 and -80 °C allowed)Other information:Avoid repeated freezing and thawing.It is recommended to aliquot the product after the first thawing.Material safety note:This product is sold for research use only. Standard Laboratory Practices should be followed when handling this material.
Catalog No. 50-153-9843