Learn More
Medchemexpress LLC Nt1-014B | 2739805-64-0 | 98.4% | 769.24 | C40H68N2O4S4 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1375005MG
Encompass_Preferred
NT1-014B is a tryptamine-derived lipidoid intended for incorporation into lipid nanoparticles (LNPs) to facilitate transport across the blood-brain barrier. It is supplied as a high-purity research chemical used to enhance brain delivery in preclinical intravenous administration and ischemic stroke studies.
  • Enables LNPs to cross the blood-brain barrier.
  • Optimized for intravenous delivery and brain targeting.
  • High purity suitable for research applications.
  • Available as a solid or DMSO solution in multiple sizes.
  • Defined molecular formula and molecular weight for formulation work.
Catalog No. 50-002-22860