Learn More
Medchemexpress LLC Nt1-014b (nt1-lipidoid) | 2739805-64-0 | 98.4% | C40H68N2O4S4 | 10MG
SDP
Supplier:  Medchemexpress LLC HY13750010MG
Encompass_Preferred
NT1-014B is a tryptamine-derived lipidoid used to formulate ionizable lipid nanoparticles (LNPs) that enable delivery across the blood-brain barrier. Supplied as a viscous, colorless to light-yellow liquid for laboratory research and formulation studies.
  • High purity: 98.4%.
  • Suitable for LNP formulation and blood-brain barrier delivery studies.
  • Viscous liquid, colorless to light yellow.
  • Good solubility in DMSO (≈150 mg/mL).
  • Intended for research use only.
Catalog No. 50-002-59003