Learn More
Medchemexpress LLC NT1-O12B | 2739805-63-9 | 95.0% | C36H60N2O4S4 | 10MG
SDP
Supplier:  Medchemexpress LLC HY13749910MG
Encompass_Preferred
NT1-O12B is a neurotransmitter-derived lipidoid designed to enhance brain delivery of cargos that are normally impermeable to the blood-brain barrier when formulated into lipid nanoparticles. The compound is an oil with high purity and documented solubility and formulation protocols for in vitro and in vivo use.
  • Enhances delivery across the blood-brain barrier.
  • Compatible with lipid nanoparticle formulation.
  • High purity (95.0%).
  • Soluble in DMSO (200 mg/mL) for in vitro applications.
  • Proven in vivo formulation protocols for suspension and clear solutions.
Catalog No. 50-002-60441