Learn More
Selleck Chemical LLC NT157 2mg 1384426-12-3
SDP
Supplier:  Selleck Chemical LLC S82282MG
Encompass_Preferred
NT157, a selective inhibitor of IRS-1/2(insulin receptor substrate), has the potential to inhibit IGF-1R and STAT3 signaling pathways in cancer cells and stroma cells of TME leading to a decrease in cancer cell survival. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-5819