Learn More
Cayman Chemical ntIoxIdnt asy KIt 96Wel
SDP
Supplier:  Cayman Chemical 70900196 WELL
Encompass_Preferred
An alkaloid with anti-inflammatory activity reduces LPS-induced TNF-a release from and NO production in RAW 264.7 macrophages at 10 30 and 100 UM reduces paw swelling in a rat model of CFA-induced arthritis at 9 and 27 mg/kg reduces colonic inflammatory cytokine levels and maintains mucosal barrier function in a mouse model of DSS-induced colitis from 25-100 mg/kg
Catalog No. 50-289-3792