Learn More
Cayman Chemical ntIpaInhydrochlorIde 10mg
SDP
Supplier:  Cayman Chemical 2670510 MG
Encompass_Preferred
A protease inhibitor; inhibits thrombokinase, plasmin, trypsin, and papain in vitro (IC50s = 20, 93, 0.26, and 0.16 μg/ml, respectively); inhibits the MNNG-induced morphological transformation of and increases frequency of chromosomal aberrations in Syrian hamster embryo cells from 6-600 μg/ml; suppresses urethan-induced formation of cleft palates and cleft lips in mice from 25-100 mg/kg
Catalog No. 50-293-4376