Learn More
Medchemexpress LLC NTRC 0066-0 | 1817791-73-3 | 99.01% | 565.71 | 5 MG
SDP
Supplier:  Medchemexpress LLC 5MGHY100024
Encompass_Preferred
NTRC 0066-0 is a selective threonine tyrosine kinase (TTK) inhibitor, demonstrating potent anti-proliferative activity against diverse cancer cell lines. With an IC50 of 0.9 nM, it is utilized in cancer research, exhibiting low selectivity entropy and a prolonged target residence time on TTK.
  • Selective threonine tyrosine kinase (TTK) inhibitor (IC50=0.9 nM).
  • Used for cancer research.
  • Exhibits low selectivity entropy.
  • Relatively long target residence time on TTK.
  • Inhibits proliferation of diverse cancer cell lines with potency similar to classic chemotherapeutic agents.
  • Demonstrates subnanomolar potency in a TTK enzyme assay.
  • Inhibits tumor growth in a mouse xenograft model of human triple-negative breast cancer (TNBC).
Catalog No. 50-003-66276