Learn More
MSC Showa Chemical Resistant Gloves, Nitrile Coated, Foam Grip, 11 3/4 in., 1 ea.
Supplier:  MSC 18309260
Encompass
These chemical-resistant gloves provide superior protection and durability for demanding industrial applications. Featuring a seamless polyester liner, these gloves are designed for comfort during extended wear. The double-dipped, microporous nitrile coating provides excellent grip in both oily and wet environments, while the full coverage design ensures comprehensive chemical resistance.
  • Chemical-resistant for professional safety
  • Double-dipped nitrile coating for enhanced durability
  • Seamless knit liner for improved comfort
  • Foam grip for secure handling in slippery conditions
  • Silicone-free composition
  • Protects against bacteria and fungi
  • Pinked cuff style
Catalog No. 50-006-88243