Learn More
TARGETMOL CHEMICALS INC NU 7026 25MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T243325MG
Encompass_Preferred
Also available in 2 mg 5 mg 10 mg 50 mg 100 mg 200 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. NU 7026 (DNA-PK Inhibitor II) is an effective DNA-PK inhibitor (IC50 0.23 uM in cell-free assays) 60-fold selective for DNA-PK than PI3K and no inhibition against both ATM and ATR. purity: 100%
Catalog No. 50-258-0814