Learn More
Medchemexpress LLC NU074381b (compound 5b) | 98.5% | 396.91 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY168529100MG
Encompass_Preferred
NU074381b is a potent S100A4 inhibitor (compound 5b) that disrupts the formation of the S100A4-NMII complex with an IC50 value of 0.48 μM. It inhibits cell proliferation and migration, and covalently inhibits S100A4 by reacting specifically with cysteine 81.
  • Potent S100A4 inhibitor
  • Disrupts S100A4-NMII complex formation
  • Inhibits cell proliferation and migration
  • Covalently inhibits S100A4 via cysteine 81
Catalog No. 50-003-90447