Learn More
Medchemexpress LLC HY-15569 5mg Medchemexpress, NU6102 CAS:444722-95-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY155695MG
Encompass_Preferred
Medchemexpress, HY-15569 5mg NU6102 CAS:444722-95-6 NU6102 is a potent CDK1 and CDK2 inhibitor with IC50s of 9.5 nM and 5.4 nM for CDK1/cyclinB and CDK2/cyclinA3, respectively. NU6102 shows selectivity for CDK1/CDK2 over CDK4 (IC50 of 1.6 μM), DYRK1A (IC50 of 0.9 μM), PDK1 (IC50 of 0.8 μM) and ROCKII (IC50 of 0.6 μM). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1740