Learn More
Medchemexpress LLC NUCC-390 | 1060524-97-1 | 98.4% | 395.54 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY11179350MG
Encompass_Preferred
NUCC-390 is a novel and selective small-molecule CXCR4 receptor agonist. It induces internalization of CXCR4 receptors and acts in an opposite way to AMD3100. NUCC-390 has been shown to promote nerve recovery of function after neurodegeneration in vivo.
  • Novel and selective small-molecule CXCR4 receptor agonist.
  • Induces internalization of CXCR4 receptors.
  • Acts in an opposite way to AMD3100.
  • Promotes nerve recovery of function after neurodegeneration in vivo.
  • Boosts axonal growth in cultured cerebellar granule neurons (CGNs) via CXCR4.
  • Contributes to the functional and anatomical recovery of the neuromuscular junction (NMJ) following acute nerve terminal damage in CD-1 mice.
Catalog No. 50-004-39037