Learn More
Medchemexpress LLC NUCC-390 dihydrochloride | 2749281-71-6 | 98.2% | 468.46 | 100 MG
Supplier: Medchemexpress LLC HY111793A100MG
NUCC-390 dihydrochloride is a novel and selective small-molecule CXCR4 receptor agonist. It induces internalization of CXCR4 receptors and acts in an opposite way to AMD3100. NUCC-390 dihydrochloride promotes nerve recovery of function after neurodegeneration in vivo.
- Novel and selective small-molecule CXCR4 receptor agonist.
- Induces internalization of CXCR4 receptors.
- Acts in an opposite way of AMD3100.
- Promotes nerve recovery of function after neurodegeneration in vivo.
- Produces strong (Ca)i response (at 10 μM).
- Stimulates signaling activity downstream of CXCR4 receptors.
- Boosts axonal growth in cultured cerebellar granule neurons (CGNs) via CXCR4.
- Contributes to the functional and anatomical recovery of the neuromuscular junction (NMJ) following acute nerve terminal damage in vivo.