Learn More
TARGETMOL CHEMICALS INC NUCIFERINE 25MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T336925MG
Encompass_Preferred
Also available in 5 mg 10 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Nuciferine ((-)-Nuciferine) is an alkaloid found within the plants Nymphaea caerulea and Nelumbo nucifera. It has a profile of action associated with dopamine receptor blockade. purity: 99%
Catalog No. 50-257-6956