Learn More
Enzo Life Sciences EPIXTRACT Nuclear Protein Isolation Kit (100 Reactions)
SDP
Supplier:  Enzo Life Sciences ENZ450160100
Encompass_Preferred
Rapid, reliable method to isolate nuclear proteins in mammalian cells and tissues. EPIXTRACT Nuclear Protein Isolation Kit II provides a simple and reliable method to isolate nuclear proteins. Extracts can be used for a variety of downstream applications including western blotting, protein-DNA binding assays, nuclear enzyme assays and others requiring nuclear proteins. Assay Time: <1 hour.
Catalog No. 50-210-0555