Learn More
Macherey-Nagel NucleoBond PC 10000 (5)
Supplier:  Macherey-Nagel 740593
Encompass_Preferred
NucleoBond PC 10000 (5) 5 preps for the isolation of plasmid DNA - NucleoBond AX 10000 Columns, NucleoBond Folded Filters, buffers, RNase A
Catalog No. NC0389398