Learn More
Macherey-Nagel NucleoBond PC 2000 EF (5)
Supplier:  Macherey-Nagel 740549
Encompass_Preferred
NucleoBond PC 2000 EF (5) 5 preps for the isolation of endotoxin-free plasmid DNA - NucleoBond AX 2000 Columns, NucleoBond Bottle Top Filters Type 1, buffers, RNase A
Catalog No. NC0389394