Learn More
Medchemexpress LLC Nucleophosmin/Npm1 protein | 96.0% | 1 MG

Supplier: Medchemexpress LLC HYP74680A1MG
Recombinant human nucleophosmin (Npm1) protein, 193 amino acids, expressed in E. coli with an N-terminal His tag. Supplied as a lyophilized powder formulated in PBS (pH 7.4), it is intended for biochemical, structural, and antibody validation applications.
- Recombinant human sequence, 193 amino acids.
- N-terminal His tag facilitates purification and detection.
- Lyophilized formulation enables long-term storage and convenient reconstitution.
- Low endotoxin level (<1 EU/μg) suitable for sensitive assays.
- Recommended storage at -20°C; stable at 4°C short-term after reconstitution.
- Amino acid sequence provided to support experimental design.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.