Learn More
Macherey-Nagel HPLC column EC 125/4 NUCLEOSIL 120-3 C8
Supplier:  Macherey-Nagel 720071.40
Encompass_Preferred
EC HPLC column - EC 125/4 NUCLEOSIL 120-3 C8 - Length: 125 mm, ID: 4 mm - pack of 1
Catalog No. NC1431373