Learn More
Macherey-Nagel NucleoSpin miRNA, 50 preps
Supplier:  Macherey-Nagel 740971.50
Encompass_Preferred
NucleoSpin miRNA (50) 50 preps for the isolation of small and large RNA - NucleoSpin RNA Columns, NucleoSpin miRNA Columns, NucleoSpin Filters, NucleoSpin Protein Removal Columns, Collection Tubes, buffers, RNase-free rDNase
Catalog No. NC0389433