Learn More
LIFE TECH (BID ECHO ONLY) NuPAGE™ Antioxidant
Supplier:  LIFE TECH (BID ECHO ONLY) NP0005
Encompass
Add to running buffer in the upper (cathode) buffer chamber when performing protein gel electrophoresis under reducing conditions.
  • Use with samples reduced using NuPAGE Sample Reducing Agent or other reducing agents
  • Optimized for neutral pH of NuPAGE Novex™ gels and is not compatible with other gel chemistries
  • May also be added to NuPAGE Transfer Buffer to enhance transfer of proteins to membranes
  • Migrates with reduced proteins to maintain reducing conditions and to prevent reoxidation of sensitive amino acids such as methionine and tryptophan
1D Gel Electrophoresis, Protein Gel Electrophoresis, Proteins, Expression, Isolation and Analysis
Catalog No. NC3859463
Only null left
Edge