Learn More
Cayman Chemical nuropeptIde EIratrIflu 5mg
SDP
Supplier:  Cayman Chemical 414575 MG
Encompass_Preferred
An endogenous peptide fragment of rat prepro-MCH; expressed in both the central and peripheral nervous systems, localizes to the cytoplasm, and colocalizes with MCH in the CNS; reduces TRH release in mediobasal hypothalamus explants at 1 µM; increases serum levels of LH in male rats and in chronically ovariectomized female rats that received estradiol benzoate and progesterone in addition to NEI; induces excessive grooming behavior and increases locomotor activity in rats at 1 UG/animal; increases the amount of time spent in REM sleep in rats when injected intracerebroventricularly or into the vlPAG
Catalog No. 50-294-2673