Learn More
Nusep Inc NUSEP NC MEM 0.2UM 8X8 15/PK
Supplier:  Nusep Inc DBMNIM03
Encompass_Preferred
NuSep’s Nitrocellulose Transfer Membrane is a practical choice for protein research and Western blotting. It features a 0.2µm pore size and comes in a convenient 8cm x 8cm format, with 15 sheets per pack to support regular lab work.This membrane is designed for reliable protein binding, providing strong retention and clear results with minimal background interference. The uniform surface and consistent thickness ensure effective transfers, helping researchers achieve dependable results every time.Key features:-High Protein Binding: 0.2µm pore size captures proteins efficiently.-Low Background: Helps maintain clarity in results.-Pre-Cut Size: Ready-to-use 8cm x 8cm sheets save preparation time.-Durable: Sturdy for repeated handling and compatible with multiple labeling procedures.NuSep’s nitrocellulose membrane is a practical addition to any lab, providing consistency and quality for essential research tasks.
Catalog No. NC1854221