Learn More
Trajan Scientific VICI Valco Tube End Nut Fitting, Flangeless, With CTFE Ferrule, 1/4 -28, 1/8"" OD, Assorted 12 Pack, one of each Color
Supplier:  Trajan Scientific CFL2A
Encompass
VICI Valco Tube End Fitting, Flangeless, With CTFE Ferrule, 1/8" OD, Assorted 12 Pack, one of each Color. Made in the USA.
Catalog No. 50-109-0308