Learn More
IDEX Health & Science Upchurch NUT MALE 1/8IN POLY
Supplier:  IDEX Health & Science Upchurch P320
Encompass_Preferred
Idex Flangeless Nuts: Compatible with 1/4-28, Polypropylene, Certifications/Compliance: ROHS, Natural, Dimensions: 0.87 x 0.37 in. (2.2 x 9.4 cm), For Use With: For 0.13 in. O.D. Tubing, Length: 0.87 in., 2.2 cm, Length Thread: 0.48 in., 1.22 cm, Max. Pressure: 500 psi.
Catalog No. NC1919060