Learn More
IDEX Health & Science Upchurch NUT PEEK F/3/16 FLF 5/16-24
Supplier:  IDEX Health & Science Upchurch F132
Encompass_Preferred
Idex PEEK MicroTight Ferrules, For Use With: 0.15 in. O.D Tubing Connect Nut, Micro Fingertight Ferrule, Length: 1.29 cm, Width: 0.156 in., 0.396 cm
Catalog No. NC1919744