Learn More
Cayman Chemical (±)-Nutlin-3, 548472-68-0, 1 mg
SDP
Supplier:  Cayman Chemical 100043721
Encompass_Preferred
Molecular Formula C30H30Cl2N4O4, Purity ≥98%, Formula Weight 581.5, Synonym: (±)-4-[4,5-bis-(4-chlorophenyl)-2-(2-isopropoxy-4-methoxy-phenyl)-4,5-dihydro-imidazole-1-carbonyl]-piperazin-2-one; An inhibitor of p53-Mdm2 interaction (IC50 = 0.09 M); induces the expression of p53-regulated genes and exhibits potent antiproliferative activity in cells with functional p53.
Catalog No. NC0147522