Learn More
Abcam (+)-Nutlin-3, p53-MDM2 binding inhibitor, 25MG
Supplier: Abcam AB14442625MG
MW 581.5 Da, Purity >98%. p53-MDM2 binding inhibitor (IC₅₀ = 13 μM). Less potent enantiomer of (-)-Nutlin-3 (Asc-4428
The product is subject to the following: Abcam Restricted Use Statement